Learn More
Cole-Parmer Memmert UF 55 PLUS 115 VOLT Universal Mechanical Oven, Twin Display, 1.9 Cuft, 115V DFS Item
Manufacturer:  Cole-Parmer 5220029
These mechanical convection ovens are perfect for heating and drying applications in research or industrial environments. Featuring the Memmert ControlCOCKPIT for quick and intuitive control with simple touch, turn and go operation. Durable construction with a scratch-resistant structured stainless steel exterior and easy-to-clean corrosion-resistant stainless steel interior. Recessed grips make oven easy to carry and handle and stainless steel door can be changed from left to right side opening. Other features include an internal data logger with a storage capacity of 10 years and an Ethernet interface for uploading and implementing programs and for online logging.
Catalog No. NC1538165