Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEN1 Mouse anti-Human, Clone: 7D3E10, Abnova™

Mouse monoclonal antibody raised against partial recombinant human MEN1.

Manufacturer:  Abnova Corporation MAB12246

Catalog No. 89-934-538


Add to cart

Description

Description

This gene encodes menin, a putative tumor suppressor associated with a syndrome known as multiple endocrine neoplasia type 1. In vitro studies have shown menin is localized to the nucleus, possesses two functional nuclear localization signals, and inhibits transcriptional activation by JunD, however, the function of this protein is not known. Two messages have been detected on northern blots but the larger message has not been characterized. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Specifications

Specifications

MEN1
7D3E10
Mouse monoclonal antibody raised against partial recombinant human MEN1.
In PBS (0.05% sodium azide)
MEAI/SCG2
Mouse
IgG1
RUO
Primary
4221
ELISA, Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA (1:10000) Flow Cytometry (1:200-1:400) Immunohistochemistry (1:200-1:1000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Liquid
MEN1
Recombinant protein corresponding to amino acids 392-554 of human MEN1.
100 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit