Learn More
MEN1 Rabbit anti-Human, Unconjugated, Clone: ACBA-13, Abnova™
Rabbit Monoclonal Antibody.
Manufacturer:  Abnova Corporation MAB22018
Catalog No. 89-339-756
Add to cart
Description
Description
MEN1 Monoclonal antibody specifically detects MEN1 in Human samples. It is validated for IP, ICC, WB-Ce, IF.
Specifications
Specifications
MEN1
ACBA-13
multiple endocrine neoplasia I
Liquid
Rabbit
IgG
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence
Unconjugated
MEN1
MEAI, SCG2
A synthetic peptide corresponding to human MEN1
100 μg
Primary
4221