Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mena, Mouse anti-Human, Clone: ENAH/1988, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00 - $506.50
Specifications
Antigen | Mena |
---|---|
Clone | ENAH/1988 |
Concentration | 0.2 mg/mL |
Dilution | Western Blot 1-2 ug/ml, ELISA, Protein Array |
Applications | Western Blot, ELISA, Peptide Array |
Description
Mena Monoclonal specifically detects Mena in Human samples. It is validated for Western Blot, ELISA, Protein Array.Specifications
Mena | |
0.2 mg/mL | |
Western Blot, ELISA, Peptide Array | |
Unconjugated | |
Mouse | |
Cancer, Neuroscience | |
10 mM PBS with 0.05% BSA | |
ENA, enabled homolog (Drosophila), FLJ10773, MENANDPP1, protein enabled homolog | |
ENAH | |
IgG2c κ | |
Protein A or G purified |
ENAH/1988 | |
Western Blot 1-2 ug/ml, ELISA, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Q8N8S7, Q8N8S7 | |
55740 | |
Recombinant fragment of human Mena protein (around aa 485-589) (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title