Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Menin Mouse anti-Human, Alexa Fluor 700, Clone: 7D3E10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237251AF700

Catalog No. NBP237251X

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Menin
7D3E10
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Purified
MEN1
Purified recombinant fragment of human MEN1 (AA: 392-554) expressed in E. Coli.
Protein A or G purified
Cancer, DNA Repair, Transcription Factors and Regulators, Tumor Suppressors
Primary
4221
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
menin, multiple, multiple endocrine neoplasia I
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit