Learn More
MEP1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 246401AP
Catalog No. 50-173-4466
Add to cart
Description
Description
MEP1A (Meprin A Subunit Alpha) is a Protein Coding gene. Among its related pathways are Collagen chain trimerization. Gene Ontology (GO) annotations related to this gene include metalloendopeptidase activity and metallopeptidase activity.
Specifications
Specifications
MEP1A
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P28825, Q16819, Q64230
Endopeptidase 2, MEP1A, Meprin A subunit alpha, PABA peptide hydrolase, PPH alpha, PPHA
Rabbit
IgG
150 μL
-20°C
Primary
17287, 25684, 4224
Western Blot
Unconjugated
MEP1A
Liquid
Mep1a
MEP1A Fusion Protein Ag18950
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat