Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Meprin alpha Subunit/MEP1A Rabbit anti-Human, Clone: 17, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | Meprin alpha Subunit/MEP1A |
---|---|
Clone | 17 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
Meprin alpha Subunit/MEP1A Monoclonal specifically detects Meprin alpha Subunit/MEP1A in Human samples. It is validated for ELISA.Specifications
Meprin alpha Subunit/MEP1A | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
4224 | |
IgG | |
Protein A purified |
17 | |
ELISA | |
Unconjugated | |
Rabbit | |
Human | |
bA268F1.1 (meprin A alpha (PABA peptide hydrolase)), EC 3.4.24, EC 3.4.24.18, endopeptidase-2, meprin A subunit alpha, meprin A, alpha (PABA peptide hydrolase), N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha, PABA peptide hydrolase, PPH alpha, PPHA | |
Recombinant Human Meprin alpha Subunit/MEP1A protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title