Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meprin alpha Subunit/MEP1A Rabbit anti-Mouse, FITC, Clone: 1, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP290383F

Catalog No. NB039057


Add to cart

Description

Description

Meprin alpha Subunit/MEP1A Monoclonal antibody specifically detects Meprin alpha Subunit/MEP1A in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

Meprin alpha Subunit/MEP1A
1
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Monoclonal
Mouse
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
FITC
PBS
bA268F1.1 (meprin A alpha (PABA peptide hydrolase)), EC 3.4.24, EC 3.4.24.18, endopeptidase-2, meprin A subunit alpha, meprin A, alpha (PABA peptide hydrolase), N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha, PABA peptide hydrolase, PPH alpha, PPHA
Recombinant Mouse Meprin alpha Subunit/MEP1A Protein
Protein A purified
RUO
Primary
4224
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit