Thermo Scientific™ MEPS™ Syringe Needle & Plunger for CTC Analytics Available on GSA/VA Contract for Federal Government customers only.
Save hours in sample preparation, and go from extraction to injection in a single process with Thermo Scientific™ HyperSep™ MEPS™ Syringes and Components.
Manufacturer: Thermo Scientific™ 60308105
 View more versions of this product
Catalog No. 60-308-105
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    For Use With (Equipment) CTC Analytics System
    Type Removable Needle Syringe
    For Use With CTC Analytics System
    Item Description 250uL removable needle syringe; CTC
    Save hours in sample preparation, and go from extraction to injection in a single process with Thermo Scientific™ HyperSep™ MEPS™ (Micro Extraction by Packed Sorbent) Syringe Needle & Plunger for CTC Analytics technology.
    MEPS Syringes and Components are suitable for reversed phases, normal phases, mixed mode or ion exchange chemistries and available in a variety of SPE phase chemistries. The HyperSep MEPS Barrel Insert and Needle Assembly (BIN) contains the stationary phase, and is built into the syringe needle.
    HyperSep MEPS syringes and plunger assemblies operate exactly like conventional syringes and are easily replaceable.
    MEPS performs the same functions as SPE—the removal of interfering matrix components and the selective isolation and concentration of analytes—except that it miniaturizes conventional SPE packed bed devices from milliliter bed volumes to microliter volumes, and incorporates them right into the syringe.
    Mechanically robust, the technique offers significant advantages over conventional SPE extraction for method development and other GC and LC applications:
    Significantly reduces the time to prepare and inject samples from hours to minutes—typically less than two minutes per sample
    Eliminates all extra steps between sample preparation and sample injection; the technique is fully compatible with direct injection onto LC or GC
    Reduces buffer and solvent volume from milliliters to microliters
    Reduces the required sample volume to as little as 3.6µL, making the technique especially well-suited for on-line use with LC-MS analysis of volume-limited samples
    Can easily be automated; the sample processing, extraction and injection steps are performed on-line using the same syringe (compatible with CTC and eVol platforms)
  • Description & Specifications
Related Products
Product Content Feedback