Learn More
Mer Mouse anti-Human, Alexa Fluor 350, Clone: 125518, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB8912U100UG
Catalog No. FAB8912U100
Add to cart
This item is not returnable. View return policy
Description
Description
Mer Monoclonal antibody specifically detects Mer in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Mer
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
AAB60430
MERTK
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Mer Met1-Ala499 Accession # AAB60430
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
10461
Human
Flow Cytometry
125518
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
c-Eyk, C-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MerTK, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer
Mouse
IgG2b
100 ug
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human Mer in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) Axl, rhDtk, or recombinant mouse Mer is observed.
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)