Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mer Rat anti-Mouse, Alexa Fluor 405, Clone: 3D07, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB11094148AF405

Catalog No. NBT94148405


Add to cart

Description

Description

Mer Monoclonal antibody specifically detects Mer in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Mer
3D07
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
MERTK
Recombinant Mouse Mer extracellular domain
Protein G purified
Cancer, Phospho Specific, Protein Kinase, Signal Transduction, Vision
Primary
10461
Mouse
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
c-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer
Rat
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Mouse Mer. It has no crossreactivity with rmAxl or rmDtk.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit