Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mer Antibody (3D07), Alexa Fluor™ 700, Novus Biologicals™
SDP

Rat Monoclonal Antibody

Supplier:  Novus Biologicals NB11094148AF700

Catalog No. NBT94148700


Add to Cart

Description

Description

Mer Monoclonal specifically detects Mer in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Mer
Monoclonal
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
MERTK
Recombinant Mouse Mer extracellular domain
0.1 mL
Primary
It has no cross-reactivity with recombinant mouse Axl or recombinant mouse Dtk.
Store at 4C in the dark.
IgG1
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
3D07
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
c-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer
Rat
Protein G purified
Cancer, Phospho Specific, Protein Kinase, Signal Transduction, Vision
10461
Mouse
Purified
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit