Learn More
Mer Rat anti-Mouse, Brilliant Violet 421, Clone: 108928, BD OptiBuild™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 747837
Catalog No. BDB747837
Add to cart
Description
Description
Mer Monoclonal antibody specifically detects Mer in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
Mer
108928
Brilliant Violet 421
Rat
50 μg
Refrigerate (2 to 8°C)
Monoclonal
Mouse
Flow Cytometry
0.2 mg/mL
Conjugated, Purified
IgG2a
RUO
Primary
Mer