Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mer Rat anti-Mouse, Clone: 108928, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications

$128.24 - $389.00

Specifications

Antigen Mer
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Format Purified
Clone 108928
Applications Flow Cytometry, CyTOF
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB5912SP
View Documents
R&D Systems
MAB5912SP
25 ug
Each for $128.24
Add to cart
 
MAB5912100
View Documents
R&D Systems
MAB5912100
100 ug
Each for $389.00
Add to cart
 
Description

Description

Mer Monoclonal specifically detects Mer in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications

Specifications

Mer
Purified
Flow Cytometry, CyTOF
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
108928
Unconjugated
IgG2a
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit