Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mer Rat anti-Mouse, Clone: 3D07, Novus Biologicals™
Rat Monoclonal Antibody
$177.16 - $493.00
Specifications
Antigen | Mer |
---|---|
Clone | 3D07 |
Concentration | LYOPH |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11094148
|
Novus Biologicals
NB11094148 |
0.1 mg |
Each for $493.00
|
|
NB018056
|
Novus Biologicals
NB110941480.025MG |
0.025 mg |
Each for $177.16
|
|
Description
Mer Monoclonal specifically detects Mer in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Mer | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
c-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer | |
MERTK | |
IgG1 | |
Protein G purified |
3D07 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rat | |
Cancer, Protein Kinase, Signal Transduction, Vision | |
Q60805 | |
10461 | |
Recombinant Mouse Mer extracellular domain | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title