Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mer Antibody (3D07), DyLight 680, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NB11094148FR
Description
Mer Monoclonal specifically detects Mer in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Mer | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MERTK | |
Recombinant Mouse Mer extracellular domain | |
0.1 mL | |
Primary | |
It has no cross-reactivity with recombinant mouse Axl or recombinant mouse Dtk. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
3D07 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
c-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer | |
Rat | |
Protein G purified | |
Cancer, Phospho Specific, Protein Kinase, Signal Transduction, Vision | |
10461 | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction