Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercaptoacetic acid, 97+%, Thermo Scientific Chemicals

Catalog Number AAB203910E
Click to view available options
Quantity:
100 g
500 g
2500 g
68-11-1
C2H4O2S
92.112
MFCD00004876
CWERGRDVMFNCDR-UHFFFAOYSA-N
mercaptoacetic acid, thioglycolic acid, 2-thioglycolic acid, acetic acid, mercapto, 2-mercaptoacetic acid, sulfanylacetic acid, thiovanic acid, mercaptoessigsaeure, thioglycollic acid, glycolic acid, thio
1133
CHEBI:30065
2-sulfanylacetic acid
C(C(=O)O)S
68-11-1
C2H4O2S
92.112
MFCD00004876
CWERGRDVMFNCDR-UHFFFAOYSA-N
mercaptoacetic acid, thioglycolic acid, 2-thioglycolic acid, acetic acid, mercapto, 2-mercaptoacetic acid, sulfanylacetic acid, thiovanic acid, mercaptoessigsaeure, thioglycollic acid, glycolic acid, thio
1133
CHEBI:30065
2-sulfanylacetic acid
C(C(=O)O)S

Air Senstitive

Reagent that protects tryptophan in amino acid analysis.Mercaptoacetic acid is used as a protecting agent for tryptophan in amino acid analysis and an acidity indicator. It finds application as an intermediate in the chemical reactions such as addition, elimination and cyclization. It acts as a precursor to ammonium thioglycolate, sodium thioglycolate and calcium thioglycolate. Its organotin derivatives are used as stabilizers for polyvinyl chloride (PVC). In organic synthesis, it acts as a nucleophile in thioglycolysis reactions and sulfur transfer agent for sulfonyl chloride synthesis. Further, it is used in leather processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent that protects tryptophan in amino acid analysis.Mercaptoacetic acid is used as a protecting agent for tryptophan in amino acid analysis and an acidity indicator. It finds application as an intermediate in the chemical reactions such as addition, elimination and cyclization. It acts as a precursor to ammonium thioglycolate, sodium thioglycolate and calcium thioglycolate. Its organotin derivatives are used as stabilizers for polyvinyl chloride (PVC). In organic synthesis, it acts as a nucleophile in thioglycolysis reactions and sulfur transfer agent for sulfonyl chloride synthesis. Further, it is used in leather processing.

Solubility
Miscible with water, ethanol, ethers, ketones, esters, chlorinated hydrocarbons, benzene and aromatic hydrocarbons. Slightly miscible with chloroform.

Notes
Air sensitive. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY
Melting Point -12°C
Density 1.295
Boiling Point 220°C
Flash Point 119°C (246°F)
Odor Stench
Refractive Index 1.5045
Linear Formula HSCH2CO2H
Quantity 2500 g
UN Number UN1940
Beilstein 506166
Sensitivity Air Sensitive
Merck Index 14,9336
Solubility Information Miscible with water,ethanol,ethers,ketones,esters,chlorinated hydrocarbons,benzene and aromatic hydrocarbons. Slightly miscible with chloroform.
Formula Weight 92.11
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material Mercaptoacetic acid, 98%
Show More Show Less
Hazard Category H301+H311+H331-H314-H335
Hazard Statement GHS H Statement
H301-H311-H331-H314
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: THIOGLYCOLIC ACID
EINECSNumber 200-677-4
RTECSNumber AI5950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit