Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercaptoacetic Acid, 98%, ACROS Organics™

Click to view available options
Quantity:
10mL
250mL
1L
2.5L
Packaging:
Glass bottle
68-11-1
C2H4O2S
92.11
MFCD00004876
CWERGRDVMFNCDR-UHFFFAOYSA-N
mercaptoacetic acid,thioglycolic acid,2-thioglycolic acid,acetic acid, mercapto,2-mercaptoacetic acid,sulfanylacetic acid,thiovanic acid,mercaptoessigsaeure,thioglycollic acid,glycolic acid, thio
1133
CHEBI:30065
2-sulfanylacetic acid
C(C(=O)O)S
68-11-1
C2H4O2S
92.11
MFCD00004876
CWERGRDVMFNCDR-UHFFFAOYSA-N
mercaptoacetic acid,thioglycolic acid,2-thioglycolic acid,acetic acid, mercapto,2-mercaptoacetic acid,sulfanylacetic acid,thiovanic acid,mercaptoessigsaeure,thioglycollic acid,glycolic acid, thio
1133
CHEBI:30065
2-sulfanylacetic acid
C(C(=O)O)S
Color Colorless
Viscosity 6.55 Pas (20°C)
Density 1.3250g/mL
Specific Gravity 1.325
Refractive Index 1.5030 to 1.5050
Flash Point 126°C
Infrared Spectrum Authentic
Boiling Point 220.0°C
Quantity 2.5L
Melting Point -16.0°C
Packaging Glass bottle
Assay Percent Range 98%
Linear Formula HSCH2CO2H
Fieser 01,1153; 07,366
Merck Index 15, 9489
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,ether,chloroform and benzene
Formula Weight 92.11
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Mercaptoacetic acid
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger