Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Mercaptoacetic acid calcium salt trihydrate, 98%

Manufacturer:  Alfa Aesar™ 4555709

Quantity 10g

 View more versions of this product

Catalog No. AA4555709


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

65208-41-5
(HSCH2CO2)Ca·3H2O
MFCD00150707
14,1711
Mercaptoacetic acid calcium salt trihydrate
C4H6CaO4S2·3H2O
276.34 (222.29 Anhydrous)
3755791
Calcium mercaptoacetate trihydrate; Calcium thioglycolate trihydrate

Specifications

White
10g
Powder
(HSCH2CO2)Ca·3H2O
MFCD00150707
14,1711
Mercaptoacetic acid calcium salt trihydrate
280° to 290°C (dec.)
65208-41-5
C4H6CaO4S2·3H2O
276.34 (222.29 Anhydrous)
3755791
Calcium mercaptoacetate trihydrate; Calcium thioglycolate trihydrate
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit