Mercon Mercury Spill Control Kit Consumables
Refills and replacment components for Mercury spill control kit
Manufacturer: ROSS HEALTHCARE INC 20 MVAP 475
 View more versions of this product
Catalog No. 17-976-6
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) For cleaning heavy mercury concentration, decontamination, highly concentrated, liquid form, quick action, cleans/coats surfaces, penetrates cracks
    Format Liquid
    Includes Mercury Vapor Control Reagent
    Weight (English) 16 oz.
    Capacity (English) 0.12 gal.
    Capacity (Metric) 0.47L
    Product Type Spill Control Kit
  • Description & Specifications
Related Products
Product Content Feedback