Mercon Mercury Spill Control Kit Consumables
Refills and replacment components for Mercury spill control kit
Manufacturer: ROSS HEALTHCARE INC 20 MWASH
 View more versions of this product
Catalog No. 17-977
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) For cleaning mercury contaminated instruments, tools, switches, containers
    Includes MerconWASH
    Capacity (English) 1 gal.
    Capacity (Metric) 4L
    Product Type Spill Control Kit
  • Description & Specifications
Related Products
Product Content Feedback