Mercon Mercury Spill Control Kits
Manufacturer: ROSS HEALTHCARE INC 20MKIT2
 View more versions of this product
Catalog No. 17-987-417A
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Suppressing Mercury vapor, clean up and decontaminating spilled Mercury
    Includes MERCONAspirator, 250mL MERCONspray, 475mL MERCONtainer x2, MERCONvap, 70 MERCONwipes, 1 pair of safety goggles, 2 pairs nitrile gloves, 2 disposal bags, Mercury Spill pamphlet, Mercury Waste labels, cardboard case with plastic carrying handles
    Product Type Portable Mercury Spill Kit
  • Description & Specifications
Related Products
Product Content Feedback