Learn More
Mercon Mercury Spill Control Kits
Manufacturer:  ROSS HEALTHCARE INC 20MKIT3
 View more versions of this product
Catalog No. 17-987-417B
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Suppressing Mercury vapor, clean up and decontaminating spilled Mercury
MERCONAspirator, 475mL MERCONtainer, MERCONvap, 1 pair of safety goggles, 2 pairs nitrile gloves, 2 disposal bags, Mercury Spill pamphlet, Mercury Waste labels, cardboard case with plastic carrying handles
Light Duty Mercury Spill Kit