Learn More
Mercon Mercury Spill Control Kits
Manufacturer:  ROSS HEALTHCARE INC 20 MKIT 1
 View more versions of this product
Catalog No. 17-976-2
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Wall Mountable Mercury Spill Kit
Suppressing Mercury vapor, clean up and decontaminating spilled Mercury
MERCONAspirator, 250mL MERCONspray, 475mL MERCONtainer x2, MERCONvap, 70 MERCONwipes, 1 pair of safety goggles, 2 pairs nitrile gloves, 2 disposal bags, Mercury Spill pamphlet, Mercury Waste labels, wall mounting metal case