Mercon Mercury Spill Control Kits
Manufacturer: ROSS HEALTHCARE INC 20MDRUM20L
 View more versions of this product
Catalog No. 17-977-2
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Suppressing Mercury vapor, clean up and decontaminating spilled Mercury
    Includes 4L MerconVAP mercury decontamination and vapour control solution, 2-250mL MerconSPRAY for suppressing vapour during clean-up, Mercon Mercury Aspirator for picking up HG droplets, one pair safety goggles and gloves, disposal bag, scoop, brush, squeege, Mercury Spill pamphlet
    Capacity (English) 5.28 gal.
    Capacity (Metric) 20L
    Product Type Industrial Mercury Spill Kit
  • Description & Specifications
Related Products
Product Content Feedback