Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Acetate; ACS Grade, MilliporeSigma™

Catalog Number mmx03352
Click to view available options
Quantity:
250 g
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
1600-27-7
C4H6HgO4
318.68
BRMYZIKAHFEUFJ-UHFFFAOYSA-L
mercury 2+ ion acetic acid
15337
CHEBI:33211
mercury(2+);diacetate
CC(=O)[O-].CC(=O)[O-].[Hg+2]
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
1600-27-7
C4H6HgO4
318.68
BRMYZIKAHFEUFJ-UHFFFAOYSA-L
mercury 2+ ion acetic acid
15337
CHEBI:33211
mercury(2+);diacetate
CC(=O)[O-].CC(=O)[O-].[Hg+2]
Quantity 250 g
Linear Formula Hg(CH3COO)2
Formula Weight 318.68
Grade ACS/EP/Reagent
Density 3.27g/cm3 (20°C); Bulk Density: 1000 kg/m3
Chemical Name or Material Mercuric Acetate, EMSURE™, For Analysis
Recommended Storage Store without restrictions
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit