Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Acetate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
1600-27-7
C4H6HgO4
318.68
BRMYZIKAHFEUFJ-UHFFFAOYSA-L
15337
CHEBI:33211
mercury(2+);diacetate
CC(=O)[O-].CC(=O)[O-].[Hg+2]
1600-27-7
C4H6HgO4
318.68
BRMYZIKAHFEUFJ-UHFFFAOYSA-L
15337
CHEBI:33211
mercury(2+);diacetate
CC(=O)[O-].CC(=O)[O-].[Hg+2]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 318.68
Physical Form Powder
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 3.27kg
Quantity 125g
Grade Baker Analyzed™ ACS Reagent
Chemical Name or Material Mercuric Acetate