Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Chloride, 0.200 Normal, Ricca Chemical

Click to view available options
Quantity:
500mL
1L
4L
20L
Packaging:
Cubitainer
Poly natural
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
7487-94-7,7732-18-5
Cl2Hg
271.492
LWJROJCJINYWOX-UHFFFAOYSA-L
mercuric chloride,mercury ii chloride,sublimate,mercury bichloride,mercury dichloride,corrosive sublimate,mercuric bichloride,mercury perchloride,sublimat,hgcl2
24085
CHEBI:31823
dichloromercury
Cl[Hg]Cl
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
7487-94-7,7732-18-5
Cl2Hg
271.492
LWJROJCJINYWOX-UHFFFAOYSA-L
mercuric chloride,mercury ii chloride,sublimate,mercury bichloride,mercury dichloride,corrosive sublimate,mercuric bichloride,mercury perchloride,sublimat,hgcl2
24085
CHEBI:31823
dichloromercury
Cl[Hg]Cl
Concentration or Composition Notes 0.200Normal
CAS Min % 95.0,2.59
CAS Max % 99.0,2.7
Physical Form Liquid
Packaging Cubitainer
Color Colorless
Quantity 20L
Chemical Name or Material Mercuric Chloride