Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mercuric Chloride, ACS Grade, ≥ 99.5%, LabChem™
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
Specifications
Boiling Point | 302°C |
Melting Point | 227°C |
Color | White |
CAS Max % | 100 |
Physical Form | Powder |
Quantity | 100 g |
Linear Formula | HgCl2 |
UN Number | UN1624 |
Solubility Information | Soluble in water |
Formula Weight | 271.5 |
Show More |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS H Statement Fatal if swallowed or in contact with skin. Suspected of causing cancer. Suspected of damaging fertility or the unborn child. May cause damage to organs (nervous system) through prolonged or repeated exposure. Very toxic to aquatic life with long lasting effects. |
Precautionary Statement | GHS P Statement Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Do not breathe dust. Do not get in eyes, on skin, or on clothing. Do not eat, drink or smoke when using this product. Wear protective gloves, protective clothing, eye protection, face protection. Wash exposed skin thoroughly after handling. Avoid release to the environment. If exposed or concerned: Get medical advice/attention. If swallowed: Rinse mouth. Immediately call a poison center/doctor. If on skin: Wash with plenty of soap and water. Immediately call a poison center/doctor. Take off immediately all contaminated clothing and wash it before reuse. Collect spillage. Store locked up. Dispose of contents/container to comply with local, state and federal regulations. |
EINECSNumber | 231-299-8 |
Recommended Storage | Room Temperature |
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction