Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Iodide, Powder, Red, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Poly Bottle
7774-29-0
HgI2
454.401
YFDLHELOZYVNJE-UHFFFAOYSA-L
mercury diiodide,mercuric iodide,mercury ii iodide,red mercuric iodide,mercuric iodide, red,mercury biniodide,mercury iodide,mercurius bijodatus,hydrargyrum diodatum,hydrargyrum bijodatum
24485
CHEBI:49659
diiodomercury
I[Hg]I
7774-29-0
HgI2
454.401
YFDLHELOZYVNJE-UHFFFAOYSA-L
mercury diiodide,mercuric iodide,mercury ii iodide,red mercuric iodide,mercuric iodide, red,mercury biniodide,mercury iodide,mercurius bijodatus,hydrargyrum diodatum,hydrargyrum bijodatum
24485
CHEBI:49659
diiodomercury
I[Hg]I

High quality chemicals for laboratory and specialized industrial use

Formula Weight 454.4
Physical Form Powder
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 6.36kg
Color Red
Quantity 500g
Chemical Name or Material Mercuric Iodide