Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mercuric Nitrate, 0.282N, 1mL = 10mg Cl-, Ricca Chemical
Click to view available options
Quantity:
500mL
1L
4L
Specifications
Concentration or Composition (by Analyte or Components) | 0.282N |
CAS Min % | 3.32,93.0,1.71 |
CAS Max % | 3.45,97.0,1.77 |
Physical Form | Liquid |
Color | Colorless |
Quantity | 1L |
Chemical Name or Material | Mercuric Nitrate |