Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Oxide, Powder, Red, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
2.5kg
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

21908-53-2
HgO

High quality chemicals for laboratory and specialized industrial use

Formula Weight 216.59
Physical Form Powder
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 11.14kg
Color Red
Quantity 500g
Chemical Name or Material Mercuric Oxide
Grade Baker Analyzed™ ACS Reagent