Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Oxide, Red, ACS Reagent (Mercury (II) Oxide), Reagents

Catalog No. 01337894
Change view
Click to view available options
Quantity:
30 g
125 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-894 30 g
01-337-895 125 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-894 Supplier Reagents Holdings Llc Supplier No. C124700030Z
Only null left
Add to Cart
Add to Cart

Mercuric Oxide, Red, ACS Reagent (Mercury (II) Oxide), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 21908-53-2
Molecular Formula HgO
Molecular Weight (g/mol) 216.59
InChI Key UKWHYYKOEPRTIC-UHFFFAOYSA-N
Synonym Mercury(II) Oxide
IUPAC Name oxomercury
SMILES O=[Hg]

Specifications

Melting Point 500°C
Color Red to Orange/Red
Physical Form Solid
Quantity 30 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 11.14
Chemical Name or Material Mercuric Oxide, Red, ACS Reagent (Mercury (II) Oxide)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.