Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Oxide, Red, ACS Reagent (Mercury (II) Oxide), Reagents

Catalog No. p-200021227
Click to view available options
Quantity:
30 g
125 g

Mercuric Oxide, Red, ACS Reagent (Mercury (II) Oxide), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 21908-53-2
Molecular Formula HgO
Molecular Weight (g/mol) 216.59
InChI Key UKWHYYKOEPRTIC-UHFFFAOYSA-N
Synonym Mercury(II) Oxide
IUPAC Name oxomercury
SMILES O=[Hg]

Specifications

Melting Point 500°C
Color Red to Orange/Red
Physical Form Solid
Quantity 125 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 11.14
Chemical Name or Material Mercuric Oxide, Red, ACS Reagent (Mercury (II) Oxide)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.