Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Sulfate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle

Chemical Identifiers

7783-35-9
HgO4S
296.648
DOBUSJIVSSJEDA-UHFFFAOYSA-L
24544

High quality chemicals for laboratory and specialized industrial use

CAS 7783-35-9
Molecular Formula HgO4S
InChI Key DOBUSJIVSSJEDA-UHFFFAOYSA-L
SMILES [O-]S(=O)(=O)[O-].[Hg+2]
IUPAC Name mercury(2+);sulfate
Molecular Weight (g/mol) 296.648
PubChem CID 24544
Formula Weight 296.65
Packaging Poly Bottle
Quantity 500g
Chemical Name or Material Mercuric Sulfate
Grade Baker Analyzed™ ACS Reagent
Show More Show Less