Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Sulfate-Silver Nitrate Special Reagent, for Manganese Determination, Ricca Chemical

Click to view available options
Quantity:
1L
Packaging:
Glass amber
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
7732-18-5,7697-37-2,7664-38-2,7783-35-9,7761-88-8
49.0,29.22,16.43,3.65,0.0017
51.0,30.41,17.1,3.8,0.0017
Mercuric Sulfate-Silver Nitrate Special Reagent, For Manganese Determination
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
7732-18-5,7697-37-2,7664-38-2,7783-35-9,7761-88-8
49.0,29.22,16.43,3.65,0.0017
51.0,30.41,17.1,3.8,0.0017
Mercuric Sulfate-Silver Nitrate Special Reagent, For Manganese Determination
Synonym dihydrogen oxide,dihydrogen monoxide
Physical Form Liquid
Packaging Glass amber
Color Colorless
Quantity 1L