Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Thiocyanate, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
Packaging:
Wide Mouth Amber Glass Bottle
592-85-8
Hg(SCN)2
592-85-8
Hg(SCN)2

High quality chemicals for laboratory and specialized industrial use

Formula Weight 316.78
Grade BAKER ANALYZED™ Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 4.00kg
Quantity 125g
Chemical Name or Material Mercuric Thiocyanate