Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Thiocyanate Stock Solution, 4.17g/L in Methanol, APHA/EPA for Chloride, Ricca Chemical

Manufacturer:  Ricca Chemical CompanySupplier Diversity Partner 478516

Packaging Glass
Quantity 500mL

 View more versions of this product

Catalog No. 4785-16

  Check Availability
Add to cart

Alert:

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.


Specifications Description & Specifications

Chemical Identifiers

Mercuric Thiocyanate Stock Solution

Specifications

Glass
500mL
Mercuric Thiocyanate Stock Solution
APHA/EPA for Chloride
4.17g/L in Methanol
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit