Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercurous Nitrate ACS AR Crystal, Macron Fine Chemicals™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Glass
14836-60-3
Hg2(NO3)2·2H2O
14836-60-3
Hg2(NO3)2·2H2O
Formula Weight 561.22
Physical Form Crystalline
Grade ACS
Packaging Glass
Quantity 2.5kg
Chemical Name or Material Mercurous Nitrate, Dihydrate