Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury, 99.99+%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Plastic bottle
7439-97-6
Hg
200.592
QSHDDOUJBYECFT-UHFFFAOYSA-N
hydrargyrum,quicksilver,metallic,liquid silver,mercurio,quecksilber,mercure,quick silver,colloidal,mercury, metallic
23931
CHEBI:16170
mercury
[Hg]
7439-97-6
Hg
200.592
QSHDDOUJBYECFT-UHFFFAOYSA-N
hydrargyrum,quicksilver,metallic,liquid silver,mercurio,quecksilber,mercure,quick silver,colloidal,mercury, metallic
23931
CHEBI:16170
mercury
[Hg]
Quantity 5g
Cadmium (Cd) 0.1ppm max.
Copper (Cu) 0.1ppm max.
Density 13.5400g/mL
Assay Percent Range 99.99+%
Insoluble Matter 0.002% (in nitric acid) max.
Iron (Fe) 0.5ppm max.
Lead (Pb) 0.1ppm max.
Merck Index 15, 5966
Packaging Plastic bottle
Formula Weight 200.59
Physical Form Heavy Liquid
Percent Purity ≥99.990% (MB)
Silver (Ag) 1ppm max.
Zinc (Zn) 0.5ppm max.
Color Silver
Chemical Name or Material Mercury
Show More Show Less