Mercury, ACS, 99.999% (metals basis), Alfa Aesar

Maximum level of impurities: Appearance P.T., Nonvolatile matter 5ppm

Manufacturer: Alfa Aesar 00522P9

Quantity 10 lb.

 View more versions of this product

Catalog No. AA00522P9

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7439-97-6
    Physical Form Triple Distilled Liquid
    Assay Percent Range 99.999% (Metals basis)
    Molecular Formula Hg
    Formula Weight 200.59
    MDL Number MFCD00011035
    Chemical Name or Material Mercury
    Grade ACS Grade

    Specifications

    Physical Form Triple Distilled Liquid
    Formula Weight 200.59
    Quantity 10 lb.
    Color Silver
    DOT Information DOT Class: 8, Packaging Group: III
    EINECS Number 231-106-7
    Assay Percent Range 99.999% (Metals basis)
    Molecular Formula Hg
    MDL Number MFCD00011035
    CAS 7439-97-6
    Odor Odorless
    Health Hazard 1 H330-H360-H372-H400-H410: Fatal if inhaled. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
    Appearance Silver-colored liquid
    Vapor Pressure 0.00163 hPa
    Chemical Name or Material Mercury
    Grade ACS Grade

    H330-H360-H372-H400-H410: Fatal if inhaled. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit