Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury, ACS, 99.999% (metals basis), Alfa Aesar™

Click to view available options
Quantity:
0.5 lb.
1 lb.
5 lb.
10 lb.
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
7439-97-6
Hg
200.592
MFCD00011035
QSHDDOUJBYECFT-UHFFFAOYSA-N
hydrargyrum,quicksilver,metallic,liquid silver,mercurio,quecksilber,mercure,quick silver,colloidal,mercury, metallic
23931
CHEBI:16170
mercury
[Hg]
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
7439-97-6
Hg
200.592
MFCD00011035
QSHDDOUJBYECFT-UHFFFAOYSA-N
hydrargyrum,quicksilver,metallic,liquid silver,mercurio,quecksilber,mercure,quick silver,colloidal,mercury, metallic
23931
CHEBI:16170
mercury
[Hg]

Maximum level of impurities: Appearance P.T., Nonvolatile matter 5ppm

Quantity 5 lb.
Appearance Silver-colored liquid
Density 13.534g/cm3 at 20°C
Assay Percent Range 99.999% (Metals basis)
Odor Odorless
Formula Weight 200.59
Physical Form Triple Distilled Liquid
Grade ACS
Vapor Pressure 0.00163 hPa
Color Silver
Chemical Name or Material Mercury
Show More Show Less

P201-P202-P260-P264b-P270-P271-P281-P284-P304+P340-P308+P313-P310-P501c

H330-H360D-H372

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 231-106-7

RTECSNumber : OV4550000

TSCA : Yes

Recommended Storage : Ambient temperatures