Alfa Aesar™ Mercury bis(trifluoromethylsulfonyl)imide

Manufacturer: Alfa Aesar™ 4715606

Quantity 5g

 View more versions of this product

Catalog No. AA4715606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 104715-41-5
    Linear Formula [(CF3SO2)2N]2Hg
    Formula Weight 760.86
    MDL Number MFCD23380172
    UN Number UN2923
    Synonym Mercury bis(trifluoromethanesulfonyl)imide; Mercury bis(trifluoromethanesulfonimide)
    Chemical Name or Material Mercury bis(trifluoromethylsulfonyl)imide

    Specifications

    Formula Weight 760.86
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Synonym Mercury bis(trifluoromethanesulfonyl)imide; Mercury bis(trifluoromethanesulfonimide)
    Health Hazard 2 GHS H Statement
    H301-H311-H331-H314-H373
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    May cause damage to organs through prolonged or repeated exposure.
    Linear Formula [(CF3SO2)2N]2Hg
    MDL Number MFCD23380172
    CAS 104715-41-5
    UN Number UN2923
    Health Hazard 1 Danger
    Color Off-white
    Sensitivity Hygroscopic
    DOT Information Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
    Quantity 5g
    Physical Form Solid
    Chemical Name or Material Mercury bis(trifluoromethylsulfonyl)imide

    GHS H Statement
    H301-H311-H331-H314-H373
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit