Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mercury(I) chloride, 99+%, extra pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 400.0°C |
Color | White |
Physical Form | Amorphous Powder |
Quantity | 5 g |
Assay Percent Range | 99% min. (Iodimetry) |
Linear Formula | Hg2Cl2 |
Merck Index | 15, 5962 |
Solubility Information | Solubility in water: practically insoluble. Other solubilities: insoluble in ether and alcohol |
Formula Weight | 472.08 |
Percent Purity | ≥99% |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Toxic if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF SWALLOWED: Immediately call a POISON CENTER or doctor |
EINECSNumber | 233-307-5 |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction