Alfa Aesar™ Mercury(I) chloride, ACS, 99.5% min

Manufacturer: Alfa Aesar™ 3641936

Quantity 500g

 View more versions of this product

Catalog No. AA3641936

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10112-91-1
    Linear Formula Hg2Cl2
    Formula Weight 472.09
    MDL Number MFCD00011043
    UN Number UN2025
    Merck Index 14,5894
    Chemical Name or Material Mercury(I) chloride
    Grade ACS Grade

    Specifications

    Formula Weight 472.09
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Sensitivity Moisture & Light Sensitive
    Merck Index 14,5894
    Color White
    Quantity 500g
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: MERCURY COMPOUNDS, SOLID, N.O.S.
    Physical Form Powder
    Linear Formula Hg2Cl2
    MDL Number MFCD00011043
    UN Number UN2025
    Melting Point 400°C (subl.)
    Density 7.15g/mL
    TSCA TSCA
    CAS 10112-91-1
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number OV8740000
    Chemical Name or Material Mercury(I) chloride
    Assay ≤99.5%
    Grade ACS Grade

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit