Alfa Aesar™ Mercury(I) fluoride, 97+%

Manufacturer: Alfa Aesar™ 4013018

Quantity 50g

 View more versions of this product

Catalog No. AA4013018

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13967-25-4
    Formula Weight 439.18
    MDL Number MFCD00016136
    UN Number UN2025
    Chemical Name or Material Mercury(I) fluoride

    Specifications

    Formula Weight 439.18
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN2025
    Melting Point 570°C (dec.)
    Health Hazard 1 Danger
    Solubility Information Hydrolyzes in water
    Density 8.73g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY COMPOUNDS, SOLID, N.O.S.
    Physical Form Powder
    MDL Number MFCD00016136
    CAS 13967-25-4
    Quantity 50g
    Health Hazard 2 GHS H Statement
    H300-H310-H330-H373
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Odor Odorless
    Chemical Name or Material Mercury(I) fluoride
    Assay ≥97%

    GHS H Statement
    H300-H310-H330-H373
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit