Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(I) sulfate, 97%, Acros Organics

Click to view available options
Quantity:
5g
100g
Packaging:
Plastic bottle
7783-36-0
Hg2O4S
497.24
MFCD00016138
MINVSWONZWKMDC-UHFFFAOYSA-L
mercurous sulfate,dimercury sulfate,dimercury sulphate,mercury i sulfate,sulfuric acid, dimercury 1+ salt,unii-pi950n9dys,mercurous sulphate,sulfuric acid, dimercury 1++ salt,pi950n9dys
24545
mercury(1+);sulfate
[O-]S(=O)(=O)[O-].[Hg+].[Hg+]
7783-36-0
Hg2O4S
497.24
MFCD00016138
MINVSWONZWKMDC-UHFFFAOYSA-L
mercurous sulfate,dimercury sulfate,dimercury sulphate,mercury i sulfate,sulfuric acid, dimercury 1+ salt,unii-pi950n9dys,mercurous sulphate,sulfuric acid, dimercury 1++ salt,pi950n9dys
24545
mercury(1+);sulfate
[O-]S(=O)(=O)[O-].[Hg+].[Hg+]
Assay Percent Range 96.0 to 104% (Iodimetry)
Linear Formula Hg2SO4
Merck Index 15, 5965
Solubility Information Solubility in water: 0.06g/100mL (25°C). Other solubilities: soluble in diluted nitric acid
Formula Weight 497.24
Physical Form Powder
Percent Purity 97%
Packaging Plastic bottle
Solubility Solubility in water: 0.06g/100mL (25°C). Other solubilities: soluble in diluted nitric acid
Color Beige or White-Yellow
Quantity 5g
Chemical Name or Material Mercury(I) sulfate
Show More Show Less

GHS H Statement
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Fatal if swallowed.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 231-993-