Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury ICP Standard, 10 ppm in 3% HNO3, Ricca Chemical
SDP

Click to view available options
Quantity:
100mL
Packaging:
Low Density Polyethylene Bottle
7439-97-6, 7697-37-2, 7732-18-5
Hg
200.592
QSHDDOUJBYECFT-UHFFFAOYSA-N
23931
CHEBI:16170
mercury
[Hg]
7439-97-6, 7697-37-2, 7732-18-5
Hg
200.592
QSHDDOUJBYECFT-UHFFFAOYSA-N
23931
CHEBI:16170
mercury
[Hg]

ICP Standard

CAS Min % 96.89, 3.11, 0.001
CAS Max % 96.89, 3.11, 0.001
Solubility Information Miscible in water
Physical Form Liquid
Packaging Low Density Polyethylene Bottle
Density 1.02
Melting Point ∼0°C
Boiling Point ∼100°C
Color Colorless
Quantity 100mL
Chemical Name or Material Mercury ICP Standard, 10 ppm in 3% HNO3
Show More Show Less

ShelfLife : 6 months

Recommended Storage : Ambient Corrosive