Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) acetate, ACS, 98.0% min, Alfa Aesar™

Click to view available options
Quantity:
50g
250g
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
1600-27-7
C4H6HgO4
318.68
MFCD00012165
BRMYZIKAHFEUFJ-UHFFFAOYSA-L
mercury 2+ ion acetic acid
15337
CHEBI:33211
mercury(2+);diacetate
CC(=O)[O-].CC(=O)[O-].[Hg+2]
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
1600-27-7
C4H6HgO4
318.68
MFCD00012165
BRMYZIKAHFEUFJ-UHFFFAOYSA-L
mercury 2+ ion acetic acid
15337
CHEBI:33211
mercury(2+);diacetate
CC(=O)[O-].CC(=O)[O-].[Hg+2]
Assay Percent Range 98.0% min.
Beilstein 3563831
Appearance White crystals
Solubility Information Highly soluble in water. Soluble in alcohol and diethyl ether.
Formula Weight 318.68
Physical Form Crystalline
Grade ACS
Density 3.28 g/cm3 at 20°C
Odor Weak
Sensitivity Light sensitive
Color Red
Melting Point 179°C to 182°C
Quantity 50g
Chemical Name or Material Mercury(II) acetate
Show More Show Less

P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P312-P314-P330-P361-P363-P501c

H300+H310+H330-H336-H373

DOTInformation : DOT Class: 6.1, Packaging Group: II

EINECSNumber : 216-491-1

RTECSNumber : AI8575000

TSCA : Yes

Recommended Storage : Ambient temperatures