Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) chloride, 99.5%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
1kg
Packaging:
Plastic bottle
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
7487-94-7
Cl2Hg
271.5
MFCD00011041
LWJROJCJINYWOX-UHFFFAOYSA-L
mercuric chloride,mercury ii chloride,sublimate,mercury bichloride,mercury dichloride,corrosive sublimate,mercuric bichloride,mercury perchloride,sublimat,hgcl2
24085
CHEBI:31823
dichloromercury
Cl[Hg]Cl
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
7487-94-7
Cl2Hg
271.5
MFCD00011041
LWJROJCJINYWOX-UHFFFAOYSA-L
mercuric chloride,mercury ii chloride,sublimate,mercury bichloride,mercury dichloride,corrosive sublimate,mercuric bichloride,mercury perchloride,sublimat,hgcl2
24085
CHEBI:31823
dichloromercury
Cl[Hg]Cl
Assay Percent Range 99.5%
Linear Formula HgCl2
Fieser 01,652; 05,427; 06,359; 09,291; 11,467; 12,304; 13,175; 15,200
Merck Index 15, 5943
Solubility Information Solubility in water: 7.4g/100 mL (20°C). Other solubilities: 1g/40 mL acetic acid,1g/200 mL benzene- 1g/3.8 mL alcohol,1g/22 mL ether- 1g/12 mL glycerol,soluble in acetone and ethyl acetate,slightly soluble in pyridine and cs2
Formula Weight 271.5
Physical Form Crystals or Crystalline Powder
Percent Purity 99.5%
Density 5.44
Packaging Plastic bottle
Solubility Solubility in water: 7.4 g/100 mL (20°C). Other solubilities: 1g/40 mL acetic acid, 1g/200 mL benzene - 1g/3.8 mL alcohol, 1g/22 mL ether - 1g/12 mL glycerol, soluble in acetone and ethyl acetate, slightly soluble in pyridine and cs2
Storage Information May contain black particles
Boiling Point 302.0°C
Color White
Melting Point 277.0°C
Quantity 100g
Chemical Name or Material Mercury(II) chloride, p.a.
Show More Show Less

GHS P Statement
Fatal if swallowed.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.
Suspected of damaging fertility.
Causes damage to organs through prolonged or repeated exposure.

GHS P Statement
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger